Please login to your online account to display your discounted pricing
PIP™ MaxiFlex™ Nylon, Micro-Foam Nitrile Grip Gloves: Dotted Palm
Breathable gloves incorporates raised micro dots to increase durability and offer more cushioning
$79.83 - $79.83
Products
Catalog Number Mfr. No. Price Quantity    
19-168-472A
protective industrial
348745S
Dozen of 12 Pair for $79.83
19-168-472D
protective industrial
348745XL
Dozen of 12 Pair for $79.83
19-168-472B
protective industrial
348745M
Dozen of 12 Pair for $79.83
19-168-472E
protective industrial
348745XXL
Dozen of 12 Each for $79.83
Description & Specifications
Offers nitrile dotted palms, black micro-foam nitrile-coated palms and fingertips, and a 15-gauge gray seamless knit nylon liner.
  • Incorporate unique Micro Dots on palm for excellent grip
  • Ultra-cool and very comfortable to wear, breathable coating keeps hands cool and dry
  • Excellent dexterity, sensitivity and tactility
  • Designed to produce minimal lint and dust
  • Excellent dry grip
  • Minimal penetration keeps the liner soft on the inside of the glove
  • Cuff style: Continuous knit
Applications:
Used in applications which require a high degree of dexterity and sensitivity, especially where grip is important, such as handling small oily parts and components, general asembly, machining, micro-engineering, automotive components, painting and horticulture.
ISO901:2000; ISO14001:2004 certified