Please login to your online account to display your discounted pricing

PIP™ Kut-Gard™ Stainless-Steel Gloves

Offer exceptional cut and slash resistance

$203.29 - $203.29

Specifications

Color Gray
Cut Resistance Rating Level 5
Glove Cuff Color Gray
Material Dyneema™
Material Polyester
View More Specs
Products
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity    

19-160-828C

 
protective industrial
22754M
Blue 9.5 in. 4 in. Medium CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

19-160-828D

 
protective industrial
22754L
White 10.5 in. 4.25 in. Large CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Gray
Cut Resistance Rating Level 5
Glove Cuff Color Gray
Material Dyneema™
Material Polyester
Material Silica
For Use With (Application) application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling
Glove Cuff Style Elastic knit
Weight Light weight

Ideal for any applications where sharp cutting instruments are used. Gloves have steel wire core with three strands of stainless steel wound together and wrapped in polyester to ensure maximum protection.

  • Yarns constructed of flexible steel core strands
  • Seamless construction provides the comfort, fit and dexterity of a knit glove
  • Gray color hides dirt

Light duty
Gauge: 13mil