missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PIP™ Kut-Gard™ Light-Duty Cut-Resistant Gloves
Ideal for any application where sharp cutting instruments are used, such as food service, food processing, and glass and metal handling.
$121.87 - $121.87
Specifications
Color | White |
---|---|
Material | Polyester,Stainless Steel |
For Use With (Application) | application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling |
Glove Cuff Style | Elastic knit |
Weight | Light weight |
Catalog Number | Mfr. No. | Length (English) | Size | Hem Color | Width (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Size | Hem Color | Width (English) | Price | Quantity | ||
19-171-211
|
PIP™
GP22730L |
10 in. | Large | White | 4.25 in. |
Case of 12 Each for $121.87
|
|
19-171-210
|
PIP™
GP22730M |
9.5 in. | Medium | Blue | 4.25 in. |
Case of 12 Each for $121.87
|
|
19-171-208
|
PIP™
GP22730XS |
8 in. | X-Small | Black | 3.5 in. |
Case of 12 Each for $121.87
|
|
19-171-209
|
PIP™
GP22730S |
8.5 in. | Small | Yellow | 3.75 in. |
Case of 12 Each for $121.87
|
|
19-171-212
|
PIP™
GP22730XL |
11 in. | X-Large | Green | 4.5 in. |
Case of 12 Each for $121.87
|
|
Description
- Seamless knit construction provides comfort, fit, and dexterity
- PolyKor™ engineered yarns offer exceptional cut and slash resistance
- Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
- Special non-latex Cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
- Can be worn on its own or as a glove liner
- Good gripping properties
- Shrink resistant
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Products constructed with PolyKor engineered yarn are suitable for protection against mechanical hazards in the toughest environments
- PolyKor products are also highly resistant to abrasion and chemicals, so you can rely on them to provide long-lasting protection
Specifications
White | |
application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling | |
Light weight |
Polyester,Stainless Steel | |
Elastic knit |