PIP™ Kut-Gard™ Seamless Knit Heavy Weight Dyneema™ Blended Gloves
Ideal for any application where sharp cutting instruments are used, such as food service, food processing, and glass and metal handling.
Manufacturer:  PIP™ 22690XS
 View more versions of this product
Catalog No. 19-130-4476A
Description
Description
  • Seamless knit construction provides comfort, fit, and dexterity
  • Dyneema™ blend enhances the gloves performance without sacrificing flexibility or dexterity and ensures maximum protection
  • Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
  • Special non-latex cuff dramatically reduces cuff blowout
  • Excellent gripping properties
  • Shrink resistant
  • Can be washed in caustic detergents and bleach
  • Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
  • Dyneema is 15 times stronger than steel on a weight for weight basis - a High Performance Polyethylene (HPPE) fiber made from Ultra-High Molecular Weight Polyethylene (UHMWPE)
  • Dyneema fiber is much thinner and more flexible than traditional materials, such as leather and cotton, while offering higher protection
Specifications
Specifications
Level 5
application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling
X-Small
White
Dyneema™, Stainless Steel Fiber, Polyester
Dyneema™, Stainless Steel, Polyester
Heavy weight
Documents