Please login to your online account to display your discounted pricing

PIP™ Kut-Gard II Spectra-Wrapped Stainless Steel Core Cut-Resistant Gloves

$294.05 - $294.05

Specifications

Color White
Cut Resistance Rating Level 5
Material Polyester
Material Stainless Steel
For Use With (Application) application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-130-4476A

 
protective industrial
22690XS
X-Small CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

19-130-4476B

 
protective industrial
22690S
Small CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

19-130-4476C

 
protective industrial
22690M
Medium CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

19-130-4476D

 
protective industrial
22690L
Large CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

19-130-4476E

 
protective industrial
22690XL
X-Large CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color White
Cut Resistance Rating Level 5
Material Polyester
Material Stainless Steel
For Use With (Application) application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling
Weight Heavy weight