missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ Kut-Gard™ Gloves

Offer exceptional cut and slash resistance

$15.72 - $15.72

Specifications

Color White
Cut Resistance Rating Level 5
Material Stainless Steel,Synthetic
For Use With (Application) application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling
Glove Cuff Style Elastic knit
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity  
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity  
19-130-4475E Protective Industrial
22600XL
Green 11 in. 4.75 in. X-Large Each for $15.72
Case of 24 Each for $366.24
19-130-4475C Protective Industrial
22600M
Blue 10 in. 4.5 in. Medium Each for $15.72
Case of 24 Each for $366.24
Specifications

Description

Gloves of steel wire core have three strands of stainless steel wound together and wrapped in polyester to ensure maximum protection.

  • Constructed of flexible steel core strands
  • Seamless construction provides the comfort, fit and dexterity of a knit glove
  • Blue color hides dirt

Description & Specifications

Specifications

White
Stainless Steel,Synthetic
Elastic knit
Level 5
application where sharp cutting instruments are used, such as food service, food processing and Glass and Metal handling
Heavy weight