Please login to your online account to display your discounted pricing

PIP™ G-Tek™ NPB2 Nylon Gloves

Seamless knits for general duty

$32.37 - $32.37

Specifications

Cleaning Procedure Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
Coated Area Palm, finger
Color Black
Material Urethane
Exterior Finish Seamless
View More Specs
Products
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity    

19175104A

 
protective industrial
33325S
White 8.5 in. 3.3 in. Small Dozen of 12 Pair for $32.37

19175104B

 
protective industrial
33325M
Green 8.9 in. 3.4 in. Medium Dozen of 12 Pair for $32.37

19175104C

 
protective industrial
33325L
Brown 9.3 in. 3.7 in. Large Dozen of 12 Pair for $32.37
Description & Specifications

Specifications

Cleaning Procedure Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
Coated Area Palm, finger
Color Black
Material Urethane
Exterior Finish Seamless
For Use With (Application) electronic and computer assembly, quality control, inspection and general assembly
Glove Cuff Style Continuous knit

  • Polyurethane coated palm and fingers for abrasion resistance
  • Black seamless knit nylon for great dexterity.
  • Medium weight nylon
  • Breathable back for comfort
  • Pair packed for ease of distribution

Used in electronic and computer assembly, quality control, inspection and general assembly