missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ G-Tek™ CR Ultra Cut-Resistant Gloves

Gray spun Dyneema™ and nylon shell

$113.17 - $1,548.59

Specifications

For Use With (Application) electronics, Glass cutting, Metal handling, food processing and meat processing
Glove Cuff Style Continuous knit
Weight Medium weight
View More Specs

Products 7
Catalog Number Mfr. No. Hem Color Size Length (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Hem Color Size Length (English) Width (English) Price Quantity  
19-151-265
View Documents
PIP™
19D655M
Yellow Medium 9.4 in. 3.7 in.
Dozen of 12 Pair for $181.73
Case of 12 Dozen for $2,116.56
 
19-151-266
View Documents
PIP™
19D655L
Brown Large 9.8 in. 3.9 in.
Dozen of 12 Pair for $181.73
Case of 12 Dozen for $2,116.56
 
19-151-268
View Documents
PIP™
19D660S
White Small 8.7 in. 3.5 in.
Dozen of 12 Pair for $113.17
Case of 12 Dozen for $1,318.20
 
19-151-267
View Documents
PIP™
19D655XL
Black X-Large 10.2 in. 4.1 in.
Dozen of 12 Pair for $181.73
Case of 12 Dozen for $2,116.56
 
19-151-269
View Documents
PIP™
19D660M
Green Medium 9.1 in. 3.7 in.
Dozen of 12 Pair for $113.17
Case of 12 Dozen for $1,318.20
 
19-151-271
View Documents
PIP™
19D660XL
Yellow X-Large 9.8 in. 4.1 in.
Dozen of 12 Pair for $113.17
Case of 12 Dozen for $1,318.20
 
19-151-270
View Documents
PIP™
19D660L
Brown Large 9.4 in. 3.9 in.
Case of 144 Pair for $1,548.59
 
Description

Description

Medium weight gloves, used for electronics, glass cutting, metal handling, food processing and meat processing.

  • Black/gray Dyneema spun shell (13 gauge)
  • Black polyurethane coated palm and knuckles
  • Washable, resistant to chemicals, water and ultraviolet light
  • Seamless construction
  • Color-coded hems for easy size identification

Compliance

EN Cut level 3

Specifications

Specifications

electronics, Glass cutting, Metal handling, food processing and meat processing
Continuous knit
Medium weight
SDS