PIP™ G-Tek™ CR Plus Gloves
Medium weight, coated gloves made with Dyneema™
Manufacturer:  PIP™ 19D650XXL
 View more versions of this product
Catalog No. 19-151-254
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Polyethylene fibers 10 to 15 times stronger than steel, more durable than polyester and 40% stronger than aramid fiber. Used for electronics, glass cutting, metal handling, food processing and meat processing.
  • Gray spun Dyneema yarn knitted shell (13 gauge) with nylon/Lycra™
  • Gray polyurethane-coated palm and finger tips
  • Washable, resistant to chemicals, water and ultraviolet light
  • Seamless construction
  • Color-coded hems for easy size identification
Compliance
EN Cut Level 3
Specifications
Specifications
Continuous knit
electronics, Glass cutting, Metal handling, food processing and meat processing
2X-Large
Gray
27 cm
Blue
Dyneema™,Lycra™,Nylon
Medium weight
10.6 in.