missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PIP™ G-Tek™ CR Plus Dyneema™ Gloves with Polyurethane Grip
Thin and flexible protection
$172.38 - $172.38
Specifications
Cleaning Procedure | Recommend washing gloves in slightly warm water not exceeding 40°C, a mild non-ionic detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C |
---|---|
Coated Area | Palm, finger |
Color | White |
Material | Polyurethane |
For Use With (Application) | electronics, Glass cutting, Metal handling, food processing and meat processing |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-151-259
|
PIP™
19D625S |
Small |
Dozen of 12 Pair for $172.38
Case of 12 Dozen for $2,007.72 |
|
19-151-260
|
PIP™
19D625M |
Medium |
Dozen of 12 Pair for $172.38
Case of 12 Dozen for $2,007.72 |
|
19-151-262
|
PIP™
19D625XL |
X-Large |
Dozen of 12 Pair for $172.38
Case of 12 Dozen for $2,007.72 |
|
19-151-261
|
PIP™
19D625L |
Large |
Dozen of 12 Pair for $172.38
Case of 12 Dozen for $2,007.72 |
|
19-151-263
|
PIP™
19D625XXL |
2X-Large |
Dozen of 12 Pair for $172.38
Case of 12 Dozen for $2,007.72 |
|
Description
- Dyneema, Nylon and Lycra™ shell provides dexterity, tactile sensitivity and cut resistance
- Polyurethane coated palm and fingertips offer abrasion resistance, puncture resistance and wet/dry grip
- Knit wrist helps prevent dirt and debris from entering glove
- Resistant to chemicals, water and ultraviolet light
- Washable
- EN Cut Level 2
- White
Specifications
Recommend washing gloves in slightly warm water not exceeding 40°C, a mild non-ionic detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C | |
White | |
electronics, Glass cutting, Metal handling, food processing and meat processing | |
Dyneema™, Nylon, Lycra™ |
Palm, finger | |
Polyurethane | |
Continuous knit |