missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ G-Tek™ CR Plus Dyneema™ Cut-Resistant Gloves with Knitted Shell

Knitted shell with nylon and Dyneema filament

$136.33 - $136.33

Specifications

Color Black,Gray
Cut Resistance Rating Level 3
Material Dyneema™,Polyester,Spandex™
For Use With (Application) electronics, Glass cutting, Metal handling, food processing and meat processing
Glove Cuff Style Continuous knit
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity  
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity  
19-165-655B Protective Industrial
19D150M
Red 9.1 in. 3.7 in. Medium N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-165-655A Protective Industrial
19D150S
Purple 8.3 in. 3.5 in. Small N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Made of polyethylene fibers 10 to 15 times stronger than steel, more durable than polyester, and 40% stronger than aramid fiber. Used for electronics, glass cutting, metal handling, food processing and meat processing.

  • Seamless construction
  • Resistant to chemicals, water and UV light
  • Black nylon/Lycra™ blend, medium weight Color-coded hems for easy size identification
  • Abrasion results: 9915 cycles

Description & Specifications

Specifications

Black,Gray
Dyneema™,Polyester,Spandex™
Continuous knit
Level 3
electronics, Glass cutting, Metal handling, food processing and meat processing
Medium weight