PIP™ Falcon™ Base10™ Single-Ply PVC Rainsuits with Detachable Hoods
Ideal for construction, municipalities, shipyards, agriculture chemical spraying and industrial plant maintenance.
Manufacturer:  PIP™ 201100X4
 View more versions of this product
Catalog No. 19-130-5529
Description
Description
  • Detachable hood with drawstring
  • Storm flap front, two flapped pockets and underarm vents
  • Adjustable snaps at wrists and ankles for better fit
  • Made with .10mm thick single ply PVC
  • Elastic waist pant for easy on and off
  • Self material collar for easy cleaning
  • Not Compliant with California Proposition 65
Specifications
Specifications
Detached
64 in.
162.6cm
PVC
2.0
56 in.
Yellow
4X-Large
Documents