PIP™ PIP™ Falcon™ Rainsuits with Bib Overalls
Jacket with detachable hood and corduroy collar
Manufacturer:  PIP™ 201250X2
 View more versions of this product
Catalog No. 19-130-5519
Description
Description
  • Adjustable snaps at wrists and ankles for better fit
  • Made with .10mm thick single ply pvc
  • Elastic waist pant for easy on and off
  • Detachable hood with drawstring
  • Storm flap front
  • Two-flapped pockets
  • Underarm vents
  • Yellow
Specifications
Specifications
Detached
59 in.
149.9cm
Yellow
2X-Large
2.0
52 in.
Elastic
PVC
Documents