Please login to your online account to display your discounted pricing

PIP™ CleanTeam™ Cotton Lisle Inspection Gloves

Ideal for inspection and electronics assembly or as liners under disposable and unsupported, liquid-proof gloves

$4.13 - $4.13

Specifications

Color White
Material Cotton
For Use With (Application) inspection and electronics assembly
View More Specs
Products
Catalog Number Mfr. No. Glove Cuff Style Weight Length (Metric) Size Price Quantity    

19-046-358

 
protective industrial
97501I
Unhemmed Light weight 22cm Women's Dozen of 12 Pair for $4.13
Case of 100 Dozen for $345.00
Description & Specifications

Specifications

Color White
Material Cotton
For Use With (Application) inspection and electronics assembly

 

  • Constructed in a two-piece, reversible design
  • Minimize contamination of products from hands
  • 100% cotton
  • Length: 9 in. (23cm)