missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ Cabaret Formal White Parade Gloves w/Plastic Dotted Palm

Used for formal occasions by police, firemen, waiters, marching bands, military and pallbearers

$617.32 - $617.32

Specifications

Color White
Material Cotton
For Use With (Application) formal occasions by police, firemen, waiters, marching bands, military and pallbearers
Glove Cuff Style Open
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-150-0721 View Documents PIP™
130100WMPDM
Medium Case of 300 Pair for $617.32
19-150-0722 View Documents PIP™
130100WMPDL
Large Case of 300 Pair for $617.32
19-150-0723 View Documents PIP™
130100WMPDXL
X-Large Case of 300 Pair for $617.32
Description

Description

  • Constructed in full fashion pattern with fourchettes and set-in thumbs for a tailored fit
  • Open cuff allows for easy on and off
  • Made from 100% cotton

Specifications

Specifications

White
Cotton
formal occasions by police, firemen, waiters, marching bands, military and pallbearers
Open