Please login to your online account to display your discounted pricing

PIP™ Cabaret Formal White Parade Gloves w/o Raised Stitching

Used for formal occasions by police, firemen, waiters, marching bands, military and pallbearers

$226.19 - $226.19

Specifications

Color White
Material Cotton
For Use With (Application) formal occasions by police, firemen, waiters, marching bands, military and pallbearers
Glove Cuff Style Open
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

19-150-0715

 
protective industrial
130100WMNZXS
8.9 in. 22.6cm X-Small Case of 300 Pair for $226.19

19-150-0718

 
protective industrial
130100WMNZL
9.7 in. 24.7cm Large Case of 300 Pair for $226.19

19-150-0716

 
protective industrial
130100WMNZS
9.2 in. 23.36cm Small Case of 300 Pair for $226.19

19-150-0717

 
protective industrial
130100WMNZM
9.4 in. 24cm Medium Case of 300 Pair for $226.19

19-150-0719

 
protective industrial
130100WMNZXL
10 in. 25cm X-Large Case of 300 Pair for $226.19
Description & Specifications

Specifications

Color White
Material Cotton
For Use With (Application) formal occasions by police, firemen, waiters, marching bands, military and pallbearers
Glove Cuff Style Open

  • Constructed in full fashion pattern with fourchettes and set-in thumbs for a tailored fit
  • Open cuff allows for easy on and off
  • Made from 100% cotton