Please login to your online account to display your discounted pricing

PROTECTIVE INDUSTRIAL PRODUCTS INC PIP™ ATG™ MaxiFlex™ Cut™ Micro-Foam Nitrile Coated Gloves

Choose a glove for precision handling in dry environments. The PIP™ ATG™ MaxiFlex™ Cut™ Micro-Foam Nitrile Coated Gloves use unique high-performance yarns and fibers developed using CutTech technology to offer cut protection.

$711.61 - $711.61

Specifications

Color Green
Exterior Finish Black micro-foam nitrile
Glove Cuff Style Continuous knit
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Size Price Quantity    

19-160-390

 
protective industrial
348743XS
20.5 x 8.2cm (8 x 3.2 in.) X-Small Case of 72 Each for $711.61
Description & Specifications

Specifications

Color Green
Exterior Finish Black micro-foam nitrile
Glove Cuff Style Continuous knit

  • Unique high performance yarns and fibers developed using CutTech technology offers cut protection
  • Seamless knit engineered yarn
  • Proprietary black micro-foam nitrile coating offers 360° breathability
  • Reinforcement between thumb and forefinger, improves cut resistance and extends glove life
  • Micro-cup finish for optimized grip
  • Minimal penetration (strike through) leaving only soft liner resting against skin
  • Component material complies with Federal regulations for food contact, 21CFR, 170-199
  • Continuous knit cuff
  • EN Cut Level 3
  • Silicone free
  • Green shell

Applications:

DIY, finishing and inspection, handling of small oily parts and components, wiring operations.

CE Marked

ANSI™ Cut Level 3.