missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ MaxiChem™ ATG™ Maxidry™ Nitrile Blend CR Gloves with Liner and Non-Slip Grip

MaxiChem™ is designed and developed for those working with chemicals requiring longer and higher levels of protection.

Manufacturer:  PIP™ 56630M

 View more versions of this product

Catalog No. 19148172




Description

Description

  • Chemical Resistance - Delivered through the ATG™ LiquiTech™ technology platform
  • Ultra Lightweight - Synthetic coating combined with an industry leading super light seamless knit liner provides excellent comfort, sensitivity, fit and dexterity
  • Tailored Cuff - Keeping the cuff close to your hand so maintaining your fingers at the tip of the glove to maximize dexterity
  • Micro-cup non-slip grip finish allows for a controlled and superior grip in oily and wet applications

Specifications

Specifications

Non-slip
Gauntlet
Nylon, Lycra™
Medium
10.9cm
ISO9001:2000, ISO14001:2004, CE
12 in.
12 x 4.3 in. (30.5 x 10.9cm)
chemical sampling, offshore Petroleum, Acids and Caustics, anodizing, Solvent based cleaning, black and green liquor processes, fertilizer and pesticide handling
Nitrile
4.3 in.
51mil
Black, Green
30.5cm
Documents