missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ Assurance™ Unsupported 22mil Medium and Heavy-weight Nitrile Gloves

Resistant to physical and chemical hazards such as cut, puncture, solvents and oil based chemicals

$110.52 - $110.52

Specifications

Certifications/Compliance ISO9001:2000, 21 CFR 170-199, FDA, CE
Color Green
Material Nitrile
For Use With (Application) chemical processing, Oil refining, Petrochemicals, food processing, aerospace, automotive degreasing, automotive assembly, painting, machining operations, Metal fabrication, furniture manufacturing and electronics
Glove Cuff Style Straight
View More Specs

Products 1
Catalog Number Mfr. No. Coating Material Size Price Quantity  
Catalog Number Mfr. No. Coating Material Size Price Quantity  
19-152-502 View Documents PIP™
50N2272GXL
Sand patch X-Large Case of 12 Each for $110.52
 
Description

Description

  • Made from component materials that comply with Federal Regulations for food contact 21CFR, 170-199
  • Unlined, chlorinated
  • Straight cuff
  • 22mil thick
  • Color: green

Model: 50-N2272G

  • Sandpatch grip
  • Overall Length: 18 in. (46cm)
Model: 50-N2250G
  • Raised diamond grip
  • Overall Length: 15 in. (38cm)

Chemical processing, oil refining and petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting

Specifications

Specifications

ISO9001:2000, 21 CFR 170-199, FDA, CE
Nitrile
Straight
Green
chemical processing, Oil refining, Petrochemicals, food processing, aerospace, automotive degreasing, automotive assembly, painting, machining operations, Metal fabrication, furniture manufacturing and electronics
22mil