Please login to your online account to display your discounted pricing

PIP™ Assurance™ Unsupported 15mil Lightweight Nitrile Gloves, Flock-Lined

Provide superior resistance to physical hazards

$23.95 - $23.95

Specifications

Length (English) 13 in.
Length (Metric) 33cm
Material Nitrile
For Use With (Application) chemical processing, Oil refining, Petrochemicals, food processing, aerospace, automotive degreasing, automotive assembly, painting, machining operations, Metal fabrication, furniture manufacturing and electronics
Thickness (English) 15mil
View More Specs
Products
Catalog Number Mfr. No. Lining Color Size Price Quantity    

19-152-508

 
protective industrial
50N160GL
Flock Green Large Dozen of 12 Each for $23.95

19-152-509

 
protective industrial
50N160GXL
Flock Green X-Large Dozen of 12 Each for $23.95
Case of 12 Dozen for $274.80
Description & Specifications

Specifications

Length (English) 13 in.
Length (Metric) 33cm
Material Nitrile
For Use With (Application) chemical processing, Oil refining, Petrochemicals, food processing, aerospace, automotive degreasing, automotive assembly, painting, machining operations, Metal fabrication, furniture manufacturing and electronics
Thickness (English) 15mil

Gloves provide resistance to cut, puncture, snag and abrasion and many chemical hazards, including solvents and oil-based chemicals.

  • Light weight construction and hand specific design provide real comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
  • Raised diamond grip pattern provides superior grip
  • Chlorinated
  • Straight cuff
  • Length: 13 in. (33cm)
  • Thickness: 15mil
  • Color: green or blue

Applications:

Light chemical applications, printing and meat and poultry processing.

Comply with FDA Regulations for Food Contact. 21CFR 170-199.