PIP™ Ambi-dex™ Regular Industrial Grade Vinyl Gloves
Ideal for food handling and processing, pharmaceuticals, laboratories and electronics
Manufacturer:  PIP™ GP64V2000PFM
Catalog No. 19-121-241B
Description
Description
  • Made from a soft virgin vinyl formulation specially compounded to deliver comfort, fit, flexibility, tactile sensitivity and performance
  • Rolled cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
  • Contains no natural rubber latex proteins
  • Translucent white color
  • Ambidextrous
  • DEHP and DOP Free
  • 4.5 mil
Compliance
Made from component materials that comply with Federal Regulations for food contact 21CFR, 170-199
Specifications
Specifications
White
Medium
9.5 in.
21 CFR 170-199
4.5 mil
Vinyl
Powder Free
24 cm
Yes