Please login to your online account to display your discounted pricing

PIP™ Ambi-dex™ Regular Industrial Grade Vinyl Gloves

Ideal for food handling and processing, pharmaceuticals, laboratories and electronics

$69.71 - $69.71

Specifications

Certifications 21 CFR 170-199
Color White
Length (English) 9.5 in.
Length (Metric) 24cm
Material Vinyl
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-121-241B

 
protective industrial
64V2000PFM
Medium Case of 1000 Each for $69.71
Description & Specifications

Specifications

Certifications 21 CFR 170-199
Color White
Length (English) 9.5 in.
Length (Metric) 24cm
Material Vinyl
Glove Cuff Style Beaded/Rolled
Grade Industrial
Hand Ambidextrous
Latex Free Yes
Powdered Powder-free

  • Made from a soft virgin vinyl formulation specially compounded to deliver comfort, fit, flexibility, tactile sensitivity and performance
  • Rolled cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
  • Contains no natural rubber latex proteins

  • Translucent white color
  • Ambidextrous
  • DEHP and DOP Free
  • 4.5 mil

Made from component materials that comply with Federal Regulations for food contact 21CFR, 170-199