missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pioglitazone Hydrochloride, TCI America™

Manufacturer:  TCI America P190125G

 View more versions of this product

Catalog No. P190125G



Specifications

Specifications

Pioglitazone Hydrochloride
≥98.0% (HPLC,N)
MFCD04975446
GHUUBYQTCDQWRA-UHFFFAOYSA-N
5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
60560
Crystal-Powder at 20°C
White
112529-15-4
C19H21ClN2O3S
5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione Hydrochloride
CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl
392.898
392.90
191°C
25g

Chemical Identifiers

112529-15-4
392.898
GHUUBYQTCDQWRA-UHFFFAOYSA-N
60560
CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl
C19H21ClN2O3S
MFCD04975446
5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione Hydrochloride
5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents