Pinchcocks, Mohr Type
Clamp rubber tubing to 1.5cm dia.
$3.60 - $3.95
Specifications
Material Nickel, Steel
Product Type Clamp Pinchcock
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
S49102A Fisher Science Education
H8610
Medium Each for $3.60
Pack of 10 Each for $35.95
S49103 Fisher Science Education
H8620EMD
Large Each for $3.95
Pack of 10 Each for $39.55
Specifications
Description
  • Single-piece, flat-jaw pinchcock features nickel-plated brass construction
  • Will not corrode or rust
Description & Specifications
Specifications
Nickel, Steel
Clamp Pinchcock