Please login to your online account to display your discounted pricing

Pinacyanol chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Pinacyanol chloride
Name Note 97%
CAS 2768-90-3
Assay 96.0 to 104.0% (on dry substance)
Assay Percent Range 97%
Molecular Formula C25H25ClN2
Formula Weight 388.94
Synonym Quinaldine Blue
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC191710050

  SDS
thermo fisher scientific
191710050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.94
Physical Form Powder
Quantity 5g
Color Olive-green to green-black
Assay 96.0 to 104.0% (on dry substance)
EINECS Number 220-457-1
TSCA TSCA
Merck Index 15, 8164
Water 8.5% max.
Synonym Quinaldine Blue
UV Spectrometry Complies
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C25H25ClN2
Chemical Name or Material Pinacyanol chloride
CAS 2768-90-3
Name Note 97%