Please login to your online account to display your discounted pricing

Pinacyanol chloride, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  191710050

 View more versions of this product

Catalog No. AC191710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 388.94
Physical Form Powder
Quantity 5g
Color Olive-green to green-black
Assay 96.0 to 104.0% (on dry substance)
EINECS Number 220-457-1
TSCA TSCA
Merck Index 15, 8164
Water 8.5% max.
Synonym Quinaldine Blue
UV Spectrometry Complies
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C25H25ClN2
Chemical Name or Material Pinacyanol chloride
CAS 2768-90-3
Name Note 97%

Specifications

Chemical Name or Material Pinacyanol chloride
Name Note 97%
CAS 2768-90-3
Assay 96.0 to 104.0% (on dry substance)
Assay Percent Range 97%
Molecular Formula C25H25ClN2
Formula Weight 388.94
Synonym Quinaldine Blue