Please login to your online account to display your discounted pricing

Pinacolborane, 97%, stabilized, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371630050

 View more versions of this product

Catalog No. AC371630050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 127.98
Assay ≥96.0%
Quantity 5g
Color Clear colorless to light yellow
Density 0.8820g/mL
Refractive Index 1.3950 to 1.4000 (20°C, 589nm)
Packaging Glass bottle
Stabilizer 0.6 to 1.4% triethylamine
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
In contact with water releases flammable gases.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Synonym 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Infrared Spectrum Authentic
Molecular Formula C6H13BO2
MDL Number MFCD00674030
CAS 25015-63-8
Chemical Name or Material Pinacolborane
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Pinacolborane
CAS 25015-63-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H13BO2
Formula Weight 127.98
MDL Number MFCD00674030
Synonym 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
In contact with water releases flammable gases.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents