Please login to your online account to display your discounted pricing

Pinacolborane, 97%, stabilized, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371630250

 View more versions of this product

Catalog No. AC371630250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 127.98
Physical Form Liquid
Quantity 25g
Color Clear colorless to light yellow
Density 0.8820g/mL
Refractive Index 1.3950 to 1.4000 (20°C, 589nm)
Packaging Glass bottle
Synonym 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Assay Percent Range 97%
Molecular Formula C6H13BO2
CAS 25015-63-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
In contact with water releases flammable gases.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Stabilizer 0.6 to 1.4% triethylamine
Infrared Spectrum Authentic
Chemical Name or Material Pinacolborane
MDL Number MFCD00674030

Specifications

Chemical Name or Material Pinacolborane
CAS 25015-63-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H13BO2
Formula Weight 127.98
MDL Number MFCD00674030
Synonym 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
In contact with water releases flammable gases.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents