missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pinacol 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 418002500

 View more versions of this product

Catalog No. AC418002500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Crystalline Low Melting Solid
White
Glass bottle
99%
99%
(CH3)2COHCOH(CH3)2
76-09-5
118.18
250g
2, 3-Dimethyl-2, 3-butanediol
Authentic
Pinacol
C6H14O2
MFCD00004462

Chemical Identifiers

76-09-5
C6H14O2
118.18
2, 3-Dimethyl-2, 3-butanediol
≥98.5%
99%
(CH3)2COHCOH(CH3)2
MFCD00004462
Pinacol, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-933-5

RTECSNumber : EK1720000

TSCA : TSCA