Learn More
Pimelic Acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131235000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Pimelic acid | |
212.0°C (10.0 mmHg) | |
98% | |
C7H12O4 | |
MFCD00004425 | |
02, IV, 2003 | |
pimelic acid, 1,5-pentanedicarboxylic acid, pileric acid, heptandioic acid, heptane-1,7-dioic acid, pimelate, 6-carboxyhexanoate, 6-carboxyhexanoic acid, 1,7-heptanedioic acid, unii-bzq96wx25f | |
WLJVNTCWHIRURA-UHFFFAOYSA-N | |
heptanedioic acid | |
385 | |
160.17 |
111-16-0 | |
Authentic | |
Glass bottle | |
HO2C(CH2)5CO2H | |
500g | |
15, 7543 | |
Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in alcohol and ether,practically insoluble in cold benzene | |
OC(=O)CCCCCC(O)=O | |
160.17 | |
CHEBI:30531 | |
98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Warning
EINECSNumber : 203-840-8
RUO â Research Use Only