Invitrogen™ PIM2 Recombinant Human Protein DFS Item
PIM2 is a proto-oncogene.
Manufacturer:  Invitrogen™ LSPV4036
 View more versions of this product
Catalog No. PV4036
Description
Description
PIM2 phosphorylates BAD, which partially inhibits its apopotopic function.
Specifications
Specifications
NP_006866
Frozen
63.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PIM Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
Pim2
Baculovirus
Human
GST Tag
Documents