Invitrogen™ PIK3CG (p110 gamma) Recombinant Human Protein DFS Item
PIK3CG (p110 gamma) is the sole class IB phosphatidylinositol 3-kinase family member.
Manufacturer:  Invitrogen™ LSPV4787
 View more versions of this product
Catalog No. PV4787
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
PIK3CG (p110 gamma), along with the class IA family members, generates the important second messenger PIP2. However, unlike the class IA enzymes which signal downstream of receptor tyrosine kinases, PIK3CG primarily signals downstream of GPCRs.
Specifications
Specifications
NP_002640.2
Frozen
130.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
PI3 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 2mM DTT, 10% Glycerol
1mg
RUO - research use only
PIK3CG (p110 gamma), PI3 Kinase gamma
Baculovirus
Human
His Tag
Documents