Invitrogen™ PIK3CD/PIK3R1 (p110 delta/p85 alpha) Recombinant Human Protein, His tag DFS Item
Phosphatidylinositol 3-kinase (PI3K) is composed of a catalytic subunit (p110δ) and a regulatory subunit (p85α in this case), which are encoded by the PIK3CD and PIK3R1 genes. PI3K phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring.
$752.00 - $8,460.00
Specifications
Host Species Human
Protein Tag His Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6451 View Documents Invitrogen™
LSPV6451
20μg Each for $752.00
PV6452 View Documents Invitrogen™
LSPV6452
100μg Each for $1,944.00
PV6453 View Documents Invitrogen™
LSPV6453
1mg Each for $8,460.00
Description
Description
Phosphatidylinositol 3-kinase (PI3K) plays a role in oncogenic transformation and is consistently expressed at a high level in AML blast cells. PI3K can play a selective role in the amplification of phosphatidylinositol (3,4,5) trisphosphate (PIP3) levels that leads to neutrophil polarization, and regulates directional migration, but not random locomotion. The PI3K signaling pathway is involved in immune responses relevant to the pathogenesis of rheumatoid arthritis and other inflammatory diseases.
  • Available in 3 convenient sizes
  • Validated in our Adapta™ and Kinase Binding assays
  • A full length protein that can help advance your research when used with validated assays
Specifications
Specifications
Human
His Tag
Yes