missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PIK3CB/PIK3R1 (p110 beta/p85 alpha) Recombinant Human Protein
PIK3CB/PIK3R1 (p110β/p85α) is a phosphoinositide-3-kinase (PI3K) comprised of a catalytic subunit, p110β, and a regulatory subunit, p85α, encoded by the PIK3CB and PIK3R1 genes respectively.
$2515.00 - $10880.00
Specifications
Accession Number | NP_852664.1/NP_006210.1 |
---|---|
For Use With (Application) | Kinase Assay, Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
Formulation | 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. |
Gene ID (Entrez) | PIK3CB, PIK3R1 |
Molecular Weight (g/mol) | 84.7kDa/123.7kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV6455
|
Invitrogen™
PV6455 |
100μg |
Each for $2,515.00
|
|
|||||
PV6456
|
Invitrogen™
PV6456 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
As with other members of the PI3K family, p110β has important regulatory roles in cellular processes including cell survival, proliferation, and differentiation. It affects growth factor signaling and survival by phosphorylating inositol lipids. PTEN-deficient cancer cells have been observed to depend on p110β signaling.
- Available in 3 convenient sizes
- Validated in our Adapta™ and Kinase Binding assays
- A full-length protein that can help advance your research when used in validated assays
Specifications
NP_852664.1/NP_006210.1 | |
50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. | |
84.7kDa/123.7kDa | |
Full Length, Recombinant, Active | |
PI3 Kinase | |
Recombinant | |
Baculovirus |
Kinase Assay, Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
PIK3CB, PIK3R1 | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
phosphatidylinositol-4/p110β/p85α/p110 beta/p85 alpha/5-bisphosphate 3-kinase | |
Human | |
His Tag |