missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PIK3C2A (PI3K-C2 alpha) Recombinant Human Protein DFS Item

The Class II PI3 kinases, including PIK3C2A, phosphorylate the 3 position of PI or PI4P.

Manufacturer:  Invitrogen™ LSPV5587

 View more versions of this product

Catalog No. PV5587



Description

Description

Though not as well studied as the Class I PI3 kinases, the Class II enyzmes have been linked to diverse, recpetor-mediated cellular processes including insulin signalling, neuronal survival, and growth factor signalling.

Specifications

Specifications

NP_002636.1
Frozen
1mg
RUO - research use only
PIK3C2A (PI3K-C2 alpha)
Baculovirus
Human
GST Tag
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
171.1kDa
Kinase proteins are shipped on dry ice and should be stored at -80°C.
Catalytic
Kinase Assay
PI3 Kinase Family
Yes
Documents