Please login to your online account to display your discounted pricing

Pifithrin-μ, Tocris

Inhibitor of p53-mitochondrial binding

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material Pifithrin-μ
Assay Percent Range >99%
Molecular Formula C8H7NO2S
Formula Weight 181.21
Synonym 2-Phenylethynesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

265310

 
r & d systems
RDS265310
10mg Each for $113.57

265350

 
r & d systems
RDS265350
50mg Each for $478.51
Description & Specifications

Specifications

Chemical Name or Material Pifithrin-μ
Formula Weight 181.21
Quantity 10mg
Molecular Formula C8H7NO2S
CAS 64984-31-2
Recommended Storage Store at +4°C
Assay Percent Range >99%
Synonym 2-Phenylethynesulfonamide
Safety and Handling

Store at +4°C