Please login to your online account to display your discounted pricing

Pifithrin-α hydrobromide, Tocris


p53 inhibitor; Also aryl hydrocarbon receptor agonist

Quantity 10mg

Manufacturer: r & d systems  RDS126710

 View more versions of this product

Catalog No. 126710

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 367.3
Quantity 10mg
Chemical Name or Material Pifithrin-α hydrobromide
Molecular Formula C16H18N2OS·HBr
Assay Percent Range >98%
Synonym 1-(4-Methylphenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzothiazolyl)ethanone hydrobromide
CAS 63208-82-2
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Pifithrin-α hydrobromide
Assay Percent Range >98%
Molecular Formula C16H18N2OS·HBr
Formula Weight 367.3
Synonym 1-(4-Methylphenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzothiazolyl)ethanone hydrobromide
Accessories

Desiccate at -20°C