Please login to your online account to display your discounted pricing

Pifithrin-α hydrobromide, Tocris

p53 inhibitor; Also aryl hydrocarbon receptor agonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material Pifithrin-α hydrobromide
Assay Percent Range >98%
Molecular Formula C16H18N2OS·HBr
Formula Weight 367.3
Synonym 1-(4-Methylphenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzothiazolyl)ethanone hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

126710

 
r & d systems
RDS126710
10mg Each for $188.60

126750

 
r & d systems
RDS126750
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 367.3
Quantity 10mg
Chemical Name or Material Pifithrin-α hydrobromide
Molecular Formula C16H18N2OS·HBr
Assay Percent Range >98%
Synonym 1-(4-Methylphenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzothiazolyl)ethanone hydrobromide
CAS 63208-82-2
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C