Please login to your online account to display your discounted pricing

Alfa Aesar™ Pifithrin-alpha, p-Nitro hydrobromide

A cell-permeable p53 inhibitor

Chemical Identifiers

Chemical Name or Material Pifithrin-alpha, p-Nitro hydrobromide
CAS 389850-21-9
Formula Weight 398.3
MDL Number MFCD00398180
Synonym 1-(4-Nitrophenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzpthiazolyl)ethanone hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64277LB0

  SDS
alfa aesar™
J64277LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 398.3
CAS 389850-21-9
Chemical Name or Material Pifithrin-alpha, p-Nitro hydrobromide
MDL Number MFCD00398180
Synonym 1-(4-Nitrophenyl)-2-(4,5,6,7-tetrahydro-2-imino-3(2H)-benzpthiazolyl)ethanone hydrobromide
Sensitivity Light Sensitive & Hygroscopic
Solubility Information Soluble in DMSO or ethanol
Color Pale brown
Quantity 5mg
Physical Form Solid
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C