Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Western Blotting Filter Papers
Pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels to PVDF, nitrocellulose, or other membranes.
$79.75 - $106.51
Products
Catalog Number Mfr. No. Description Quantity Price Quantity    
PI88600
thermo scientific™
PI88600
Pierce Western Blotting Filter Paper, 8cm x 10.5cm 100 sheets Pack of 100 Each for $98.31
PI88605
thermo scientific™
88605
Pierce Western Blotting Filter Paper, Extra Thick, 7cm x 8.4cm 50 sheets Each for $86.75
PI88610
thermo scientific™
88610
Pierce Western Blotting Filter Paper, Extra Thick, 8.5cm x 9cm 50 sheets Each for $86.75
PI88615
thermo scientific™
88615
Pierce Western Blotting Filter Paper, Extra Thick, 8cm x 13.5cm 50 sheets Each for $79.75
PI88620
thermo scientific™
88620
Pierce Western Blotting Filter Paper, Extra Thick, 20cm x 20cm 50 sheets Each for $106.51
PI84783
thermo scientific™
84783
Pierce Western Blotting Filter Paper, 7cm x 8.4cm 100 sheets Each for $94.13
PI84784
thermo scientific™
84784
Pierce Western Blotting Filter Paper, 8cm x 13.5cm 100 sheets Each for $102.54
Description & Specifications
Thermo Scientific™ Pierce Western Blotting Filter Papers are pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels (SDS-PAGE) to PVDF, nitrocellulose, or other membranes.
Sheets of blotting paper are necessary components of transfer sandwiches and cassettes that typically must be assembled for various kinds of protein or nucleic acid, gel-to-membrane, transfer protocols. High-quality Pierce Western Blotting Filter Papers are available as standard-thickness and extra-thick sheets that are pre-cut for convenience with mini gels.
Highlights:
  • High-quality – clean cotton cellulose fiber paper manufactured with additive-free ultrapure water to eliminate sources of background signal and artifacts
  • Easy to use – pre-cut filter paper sheets in several convenient sizes for use with most mini gel sizes, tank transfer cassettes and semi-dry blotters
  • Two thicknesses – choose standard-thickness paper for traditional procedures or extra-thick filter paper for high capacity blotting or to eliminate using multiple sheets
  • Validated – tested and recommmend for use with protein methods, including the Thermo Scientific Pierce Fast Semi-Dry Blotter (Part No. 88217)