missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Water, LC-MS Grade

Thermo Scientific™ Pierce Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC

Manufacturer:  Thermo Scientific™ 0085189

 View more versions of this product

Catalog No. PI85189



Description

Description

Thermo Scientific™ Pierce Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.

Additional Information and Online Ordering »

An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.

Thermo Scientific Pierce Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Pierce Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Purity and quality validated with 30 tests for use in LC/MS applications
  • 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
  • Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases

Product Details:

General properties of water

  • Molecular formula: H2
  • Molecular Weight: 18.01
  • CAS number: 7732-18-5

Pierce Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

Specifications

Specifications

32°F
212°F
4 x 1 L
Liquid
Solvent (Pure)
Clear, colorless liquid
7
0.0
LC-MS
Pierce™
SDS
Documents
Product Certifications
Promotions

Promotions