Thermo Scientific™ Pierce™ Vacuum Hydrolysis Tubes
High-quality valved glass tubes designed for heated, vacuum, acid-hydrolysis of proteins and peptides for amino acid analysis by HPLC chromatography.
$273.33 - $321.70
Specifications
Quantity 1 tube
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
PI29570 Thermo Scientific™
29570
Vacuum Hydrolysis Tube, 1mL 1 tube Each for $273.33
PI29571 Thermo Scientific™
29571
Vacuum Hydrolysis Tube, 6mL 1 tube Each for $321.70
Specifications
Description
Thermo Scientific™ Pierce Vacuum Hydrolysis Tubes are high-quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis.
Highlights:
  • Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks
Description & Specifications
Specifications
1 tube