Thermo Scientific™ Pierce™ UVP 3UV Ultraviolet Lamp
Photoactivate crosslinkers in UV-crosslinking methods with these compact, UV-lamps with 252nm, 302nm and 365nm short to long wavelength bulbs.
$1,063.68 - $1,067.69
Specifications
Quantity 1 unit
Sufficient For Crosslinking methods requiring ultraviolet light for activation
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI95034 Thermo Scientific™
95034
Pierce 3UV Lamp, 115 Volts, 60 Hz (USA) Each for $1,063.68
PI95035 Thermo Scientific™
95035
Pierce 3UV Lamp, 230 Volts, 50 Hz (Europe) Each for $1,067.69
Specifications
Description
Thermo Scientific™ Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252nm, 302nm and 365nm dial-selectable wavelength settings especially for use with photoactivatable crosslinkers and UV-crosslinking methods.
The 3UV lamp from UVP is a compact ultraviolet lamp for longwave, midrange and shortwave UV applications. The lamp's innovative design allows you to dial-in the required wavelength with just the turn of a knob. The lamp is compatible with our extensive line of photo-reactive crosslinking and label-transfer reagents, including the photo-reactive amino acids for discovering in-cell protein interactions.
Highlights:
  • Multifunctional – three lamps (254nm, 302nm, 365nm) selected by a turn of the dial
  • Efficient – unique reflector design provides maximum UV light for all applications
  • Lightweight and ergonomic – designed for hand-held or stationary use
  • Small footprint – 15.5 in. x 3 in. x 4.5 in. (394mm x 76mm x 114mm)
  • Durable – lamp features aluminum construction and scratch- resistant finish
  • Versatile – 8-watt lamps are ideal for a wide area of illumination, such as culture plates
  • Validated for crosslinking – our researchers have validated this lamp for use in crosslinking applications with diazirine and phenyl azide photoactivatable crosslinkers
Description & Specifications
Specifications
1 unit
Crosslinking methods requiring ultraviolet light for activation