Please login to your online account to display your discounted pricing
Thermo Scientific™ Pierce™ Unstained Protein Molecular Weight Marker
A mixture of seven native proteins (14.4kDa to 116kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
$107.65 - $107.65
Specifications
Description Pierce Unstained Protein Molecular Weight Marker
Detection Visible
Formulation 7 proteins from 14.4 to 116kDa in reducing sample buffer
Range Mid
Stained Unstained
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    
PI26610
thermo scientific™
26610
Each for $107.65
Description & Specifications
Specifications
Description Pierce Unstained Protein Molecular Weight Marker
Detection Visible
Formulation 7 proteins from 14.4 to 116kDa in reducing sample buffer
Range Mid
Stained Unstained
Thermo Scientific™ Unstained Protein Molecular Weight Marker is a mixture of seven native proteins (14.4kDa to 116kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
The protein MW marker is designed for accurate sizing of proteins in SDS-polyacrylamide gel electrophoresis, as well as on PVDF, nylon and nitrocellulose membranes. The protein standard produces sharp bands on protein mini gels following staining with Thermo Scientific Pierce Gel Stains or other coomassie dye or silver stains.
Highlights:
  • Size range – seven proteins spanning 14.4 to 116kDa
  • Ready-to-use – supplied in a loading buffer for direct loading on gels
  • Sharp bands – equal intensity upon coomassie or silver staining
  • Quality tested – each lot evaluated by SDS-PAGE and Western blotting
Includes:
Proteins (0.1 to 0.2mg/mL of each) in 62.5mM Tris-HCl (pH 7.0 at 25°C), 1mM EDTA, 2% SDS, 50mM DTT, 30mM NaCl, 1mM NaN3, 0.01% bromophenol blue and 50% glycerol; Component proteins: beta-galactosidase (116kDa), bovine serum albumin (66.2kDa), ovalbumin (45.0kDa), lactate dehydrogenase (35.0kDa), REase Bsp98I (25.0kDa), beta-lactoglobulin (18.4kDa), lysozyme (14.4kDa)
Recommended for:
Accurate protein sizing on SDS-polyacrylamide gels and Western blots