missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Triethylamine (TEA) Solvent

Bottles of 99.5%-pure TEA (MW 101.2, CAS 121-44-8) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

$183.88 - $183.88

Specifications

For Use With HPLC
Standard Components Triethylamine
Standard Concentration 98 to 100%
Storage 2° - 8°C
View More Specs

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI25108 Thermo Scientific™
PI25108
100g Pack for $183.88
Specifications

Description

Thermo Scientific™ Pierce Triethylamine (TEA) is a high-purity solvent that can be used in certain HPLC protocols, such as weak anion exchange to resolve certain tryptic peptides on a reverse-phase column.

Triethylamine is an ion-pairing reagent that alters selectivity in reverse-phase HPLC separations. By pairing with peptides, it effectively sharpens peaks, resulting in improved peak resolution.

Highlights:

  • >99.5% triethylamine purity, allowing sensitive peptide detection at low UV wavelengths in reverse-phase HPLC peptide separation systems Packaged in amber glass bottles with protective PTFE-lined fluorocarbon caps for reagent integrity
  • Has a low UV absorbance to provide the most sensitive detection across all wavelengths

Properties of Triethylamine

  • Alternate names TEA, Diethylethanamine Molecular formula C6H15N Molecular weight 101.19 Density 0.726g/mL

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

Description & Specifications

Specifications

HPLC
Triethylamine
98 to 100%
2° - 8°C