Thermo Scientific™ Pierce™ TPCK Trypsin and Resin
Cleave the carboxyl-side of Arg and Lys residues with trypsin that isTPCK-treated to block chymotrypsin activity; its immobilized form allows enzyme removal after digestion.
$133.75 - $141.01
Specifications
MS-Compatible No
Storage Requirements 4°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI20230 Thermo Scientific™
PI20230
Pierce Immobilized Trypsin, TPCK Treated Pack for $141.01
PI20233 Thermo Scientific™
PI20233
Pierce TPCK Trypsin Pack for $133.75
Specifications
Description
Thermo Scientific™ Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.
Highlights:
  • Immobilized TPCK Trypsin eliminates trypsin contamination in digests
  • TPCK inhibits chymotrypsin activity
Description & Specifications
Specifications
No
4°C