Learn More
Thermo Scientific™ Pierce™ Fast Western Blot Kits, SuperSignal™ West Dura, Mouse
For femtogram-level detection and Western blot development less than 1 hour after gel transfer.
Supplier: Thermo Scientific™ 35070
Description
Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate contains optimized reagents and protocol that allows femtogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.
The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other rapid Western blotting systems. These complete Pierce Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Dura Chemiluminescent Substrate. Like other Western blotting methods, the included SuperSignal West Dura Substrate produces a chemiluminescent signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.Highlights:
Fast – femtogram levels of detection in about one hour
Convenient – no expensive hardware or vacuum required; no clogging issues
Simple – optimized protocol makes Western blot analysis easier than ever
Economical – no additional consumables or extra equipment to purchase
Stable – all kit components are stable for one year stored at 4°C
Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody
Compatible with:
Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer
Specifications
Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate, Mouse | |
Chemiluminescent | |
Western Blot | |
Pierce™, SuperSignal™ | |
HRP | |
Mouse | |
10 western blots (800 sq. cm of membrane) |
Western Blot Kit | |
Liquid | |
Nitrocellulose, PVDF | |
400 mL | |
Horseradish Peroxidase | |
Femtogram-level | |
Sufficient For: 10 mini blots probed with mouse primary antibodies • Fast Western Antibody Diluent, 200 mL • Fast Western 10X Wash Buffer, 100 mL • Fast Western Mouse Optimized HRP Reagent, Dura, 10 mL • SuperSignal West Dura Luminol/Enhancer Solution, 50 mL • SuperSignal West Stable Peroxide Solution, 50 mL Store kit at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.