Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ SuperSignal West Dura Fast Western Blot Kits

For femtogram-level detection and Western blot development less than 1 hour after gel transfer.

$186.39 - $638.47

Specifications

Conjugate HRP
Detection Chemiluminescent
Formulation Multi-component kit
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    

PI35070

 
thermo scientific™
35070
Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate, Mouse Each for $637.42

PI35071

 
thermo scientific™
35071
Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate, Rabbit Each for $638.47

PI35075

 
thermo scientific™
35075
Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate, Mouse Trial Size Each for $186.39

PI35076

 
thermo scientific™
35076
Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate, Rabbit Trial Size Each for $186.39
Description & Specifications

Specifications

Conjugate HRP
Detection Chemiluminescent
Formulation Multi-component kit

Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate contains optimized reagents and protocol that allows femtogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.

The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other rapid Western blotting systems. These complete Pierce Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Dura Chemiluminescent Substrate. Like other Western blotting methods, the included SuperSignal West Dura Substrate produces a chemiluminescent signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.

Highlights:

  • Fast – femtogram levels of detection in about one hour

  • Convenient – no expensive hardware or vacuum required; no clogging issues

  • Simple – optimized protocol makes Western blot analysis easier than ever

  • Economical – no additional consumables or extra equipment to purchase

  • Stable – all kit components are stable for one year stored at 4°C

  • Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody

Compatible with:

Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer