Thermo Scientific™ Pierce™ Sulfo-SBED Biotin Label Transfer Reagent
Chemically tag purified proteins at primary amines to covalently crosslink and transfer biotinylation to protein interaction binding partners.
$456.18 - $566.57
Specifications
Chemical Reactivity Amine-to-Nonselective
Cleavable Yes
Formulation Sulfo-N-hydroxysuccinimidyl-2-(6-[biotinamido]-2-(p-azido benzamido)-hexanoamido) ethyl-1,3'-dithioproprionate; powder solid
Membrane Permeable No
Molecular Weight 879.98
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI33033 Thermo Scientific™
PI33033
10mg Pack for $456.18
PI33034 Thermo Scientific™
33034
8 x 1mg Pack of 8 Each for $566.57
Specifications
Description
Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer Reagent is a multifunctional reagent for labeling a purified protein and then covalently transferring the attached biotin tag onto specific interactors of that protein.
Ready-to-use kit eliminates the need to prepare the necessary buffers or purchase the critical reagents necessary to perform a label transfer experiment. Key reagents, Sulfo-SBED and DTT, are supplied in unique, no-weigh, single-use packaging.
Highlights:
  • Sulfo-NHS ester reacts with primary amines at pH 7-9
  • Aryl azide conjugates randomly when activated with UV light
  • Cleavable disulfide bond allows biotin label transfer
  • Discover protein interactions without radiolabeling
Includes:
No-Weigh Sulfo-SBED Label Transfer Reagent, 8 × 1mg; BupH Phosphate Buffered Saline, 1 Pk. (makes 500mL); Label Transfer Buffer (20X), 200mL (makes 4L); Streptavidin-HRP Conjugate, 0.1mg; No-Weigh Dithiothreitol (DTT), 8 × 7.7mg; Slide-A-Lyzer MINI Dialysis Units plus Float, 10K MWCO, 10-100μL, 10 units/Pk.
Description & Specifications
Specifications
Amine-to-Nonselective
Sulfo-N-hydroxysuccinimidyl-2-(6-[biotinamido]-2-(p-azido benzamido)-hexanoamido) ethyl-1,3'-dithioproprionate; powder solid
879.98
14.3; 21.2; 24.7Å
Yes
No
No
Yes