Learn More
Thermo Scientific™ Pierce™ Streptavidin Agarose Columns, 1 mL
Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.
Supplier: Thermo Scientific™ LSG20351
Description
Thermo Scientific™ Pierce Streptavidin Resins comprise a wide selection of high-quality affinity beads and columns for nearly any kind of biotin-based purification procedure.
Additional Information and Online Ordering »
Highlights:
Proven coupling chemistries – the methods used to prepare these resins are superior to typical cyanogen bromide (CNBr) immobilization, providing high functionality, low nonspecific binding and excellent leach-resistance
Versatile resins – Agarose (polysaccharide-based) and UltraLink™ Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods
Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)
Superior performance – High Capacity Resin has 2 to 3 times higher binding capacity than alternative supplier products, allowing use of small amounts of resin for experiments
Item-Specific Features:
- Agarose Resins are crosslinked 6% beaded agarose
- UltraLink Resins beaded acrylamide support (UltraLink Biosupport)
- Plus and High Capacity Resins have higher loading (i.e. density) of immobilized streptavidin protein than the regular forms
Recommended for:
Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies; Cell-surface labeling with biotinylation reagents, followed by precipitation with immobilized streptavidin; Purification of cell-surface glycoproteins using biotinylated Concanavalin A; Recovery of single-stranded DNA for dideoxy sequencing
Specifications
5 mL | |
45 to 165 μm | |
5 Columns | |
Biotin-Tagged Molecule | |
Streptavidin | |
Spin Column |
Contents: 5 Ã 1 mL prepacked columns with twist-off bottom tab and accessory pack containing five white tips. Storage: Upon receipt store at 4°C. Do not freeze the resin. Product is shipped at ambient temperature. |
|
Pierce™ | |
Recombinant | |
Agarose | |
Binding 15 to 28 μg Biotin Per Column (1 to 3 mg Biotinylated BSA Per Column) | |
Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.